Sodium taurocholate hydrate 98%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: Taurocholic acid sodium salt hydrate, Cholaic acid sodium salt hydrate, Sodium 2-((R)-4-((3R,5S,7R,8R,9S,10S,12S,13R,14S,17R)-3,7,12-trihydroxy-10,13-dimethylhexadecahydro-1H-cyclopenta[a]phenanthren-17-yl)pentanamido)ethane-1-sulphonate hydrate

230370010 230370050 230370250 230371000
ACRO230370010EA 54.2 EUR
ACRO230370010 ACRO230370050 ACRO230370250 ACRO230371000
Sodium taurocholate hydrate 98%
Sodium taurocholate hydrate
Formel: C₂₆H₄₆NNaO₈S
Molecular Weight: 555.70 g/mol
Schmelzpunkt: 230 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00150819
CAS-Nummer: 345909-26-4

Bestellen

Spezifikation Testergebnisse

Appearance White to cream Powder
Infrared spectrum Conforms
Water ≤7 %
TLC ≥97.5 %
Residue after ignition 11.0 % to 14.0 %
Specific optical rotation +20.0° to +25.8° (20°C, 589 nm) (c=3, water)
Heavy metals (as Pb) ≤20 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR