4-(Aminomethyl)benzoesäure 99%

Lieferant: Thermo Fisher Scientific
Achtung

232220250 232221000
ACRO232220250EA 46.6 EUR
ACRO232220250 ACRO232221000
4-(Aminomethyl)benzoesäure 99%
4-(Aminomethyl)benzoic acid
Formel: C₈H₉NO₂
Schmelzpunkt: >300 °C
Flammpunkt: >300 °C
MDL: MFCD00010203
CAS-Nummer: 56-91-7
EINECS: 200-297-9

Bestellen

Spezifikation Testergebnisse

Appearance White to off-white Powder
Infrared spectrum Conforms
Water ≤6 % (K.F.)
HPLC ≥98.5 %
Titration with HClO4 98.5 to 101.5 % (On dry substance)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR