1-Hydroxypyrene 99+%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
232272500
232270010
232270500
ACRO232272500EA
234
EUR
ACRO232272500
ACRO232270010
ACRO232270500
1-Hydroxypyrene 99+%
1-Hydroxypyrene
CAS No.: 5315-79-7
Formel:
C₁₆H₁₀O Molecular Weight: 218.255 g/mol Schmelzpunkt: 176…178 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00044543 CAS-Nummer: 5315-79-7 |
Spezifikation Testergebnisse
Appearance | Beige to brown Powder or needles |
Infrared spectrum | Conforms |
HPLC | ≥99.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...