4-Phenylbutyrophenon 98%

Lieferant: Thermo Scientific

Synonyme: 1,4-Diphenyl-1-butanone

232820010
ACRO232820010EA 32.8 EUR
ACRO232820010
4-Phenylbutyrophenon 98%
4-Phenylbutyrophenon
Formel: (C₆H₅)C(O)CH₂CH₂CH₂(C₆H₅)
Molecular Weight: 224,3 g/mol
Siedepunkt: 155 °C (0,2 mmHg)
Schmelzpunkt: 55…59 °C
MDL: MFCD00022005
CAS-Nummer: 5407-91-0
EINECS: 226-471-4

Bestellen

Spezifikation Testergebnisse

Appearance White to light yellow Powder
Infrared spectrum Conforms
Melting point 55 to 59 °C
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR