4-Benzoylpiperidin Hydrochlorid 98%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: Phenyl(piperidin-4-yl)methanone hydrochloride

232860010 232860100
ACRO232860010EA 38.5 EUR
ACRO232860010 ACRO232860100
4-Benzoylpiperidin Hydrochlorid 98%
4-Benzoylpiperidin Hydrochlorid
Formel: C₁₂H₁₆ClNO
Schmelzpunkt: 226…227 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00066982
CAS-Nummer: 25519-80-6

Bestellen

Spezifikation Testergebnisse

Appearance White to off-white Powder
Infrared spectrum Conforms
Titration with HClO4 ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR