Prostaglandin E2 98%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
PGE₂
233560100
ACRO233560100EA
758
EUR
ACRO233560100
Prostaglandin E2 98%
Prostaglandin E2
Appearance: White to cream to light yellow Powder
Formel:
C₂₀H₃₂O₅ Molecular Weight: 352,47 g/mol Schmelzpunkt: 66…68 °C Lagertemperatur: Gefrierschrank |
MDL:
MFCD00077861 CAS-Nummer: 363-24-6 |
Spezifikation Testergebnisse
Appearance | White to cream to light yellow Powder |
Infrared spectrum | Conforms |
HPLC | ≥97.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...