Prostaglandin E1 98+%
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
Alprostadil, Caverject, PGE1, 7-((1R,2R,3R)-3-Hydroxy-2-((S,E)-3-hydroxyoct-1-en-1-yl)-5-oxocyclopentyl)heptanoic acid
235020100
ACRO235020100EA
632
EUR
ACRO235020100
Prostaglandin E1 98+%
Prostaglandin E1
Appearance: White to off-white Crystalline powder
Formel:
C₂₀H₃₄O₅ Molecular Weight: 354,487 g/mol Schmelzpunkt: 115…116 °C Lagertemperatur: Gefrierschrank |
MDL:
MFCD00077860 CAS-Nummer: 745-65-3 UN: 2811 ADR: 6.1,III |
Spezifikation Testergebnisse
Appearance | White to off-white Crystalline powder |
Infrared spectrum | Conforms |
HPLC | ≥98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...