1,2,3,4-Tetrahydro-6,7-isoquinolinediol hydrobromide 98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
235060010
ACRO235060010EA
60.1
EUR
ACRO235060010
1,2,3,4-Tetrahydro-6,7-isoquinolinediol hydrobromide 98%
1,2,3,4-Tetrahydro-6,7-isoquinolinediol hydrobromide
Appearance: Beige to grey-green to brown Powder
Formel:
C₉H₁₂BrNO₂ Molecular Weight: 246.104 g/mol Schmelzpunkt: 210 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00078115 CAS-Nummer: 52768-23-7 |
Spezifikation Testergebnisse
Appearance | Beige to grey-green to brown Powder |
Infrared spectrum | Conforms |
Titration Argentometric | ≥97.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...