Kieselgel, Porengröße: 60 Å 0.200-0.500 mm Chromatography
Lieferant: Thermo Fisher Scientific
240380010
240380050
ACRO240380010EA
91.2
EUR
ACRO240380010
ACRO240380050
Kieselgel, Porengröße: 60 Å 0.200-0.500 mm Chromatography
Silica gel
Formel:
SiO₂ Molecular Weight: 60.08 g/mol Siedepunkt: 2230 Schmelzpunkt: 1710 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00011232 CAS-Nummer: 7631-86-9 REACH: 01-2119379499-16 |
Spezifikation Testergebnisse
Appearance | White Powder |
Particle size | ≤10 % (undersized particles) |
Specific surface area | 480 to 550 m²/g |
Pore volume | 0.55 to 0.85 ml/g |
pH | 6.0 to 7.5 (5 % suspension in water) |
Residual water | ≤4 % (150°C, 2 h) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...