Phloroglucin, wasserfrei 99+%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
1,3,5-Benzoltriol, 1,3,5-Trihydroxybenzol
241761000
241760250
241765000
ACRO241761000EA
125
EUR
ACRO241761000
ACRO241760250
ACRO241765000
Phloroglucin, wasserfrei 99+%
Phloroglucin
Formel:
C₆H₆O₃ Molecular Weight: 126.11 g/mol Siedepunkt: 400 Schmelzpunkt: 215…218 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00002286 CAS-Nummer: 108-73-6 EINECS: 203-611-2 Merck Index: 12,07482 |
Spezifikation Testergebnisse
Appearance | White to light beige Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 213 to 218 °C |
Loss on drying | ≤2 % (1 g, 120 °C) |
HPLC | ≥99.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...