Phloroglucide hydrate 95%
Lieferant: Thermo Fisher Scientific
Synonyme:
2,3',4,5',6-Pentahydroxybiphenyl hydrate, PHBF
241770250
ACRO241770250EA
165
EUR
ACRO241770250
Phloroglucide hydrate 95%
Phloroglucide hydrate
CAS No.: 491-45-2
Formel:
C₁₂H₁₀O₅.xH₂O Schmelzpunkt: 300 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD01543374 CAS-Nummer: 491-45-2 |
Spezifikation Testergebnisse
Appearance | Yellow to brown Powder |
Infrared spectrum | Conforms |
HPLC | ≥94 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...