Pimozide

Lieferant: Thermo Fisher Scientific
Gefahr

242010010
ACRO242010010EA 241 EUR
ACRO242010010
Pimozide
Pimozide

  • Bioactive

Schmelzpunkt: 214…222 °C
Lagertemperatur: Kühlschrank
CAS-Nummer: 2062-78-4

Bestellen

Spezifikation Testergebnisse

Appearance White to light yellow Powder
Infrared spectrum Conforms
Melting point 214 to 222 °C
HPLC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR