Natrium 30% (w/w) Dispersion in Toluol, Ø <0.1 mm

Lieferant: Thermo Fisher Scientific
Gefahr

243262500
ACRO243262500EA 171 EUR
ACRO243262500
Natrium 30% (w/w) Dispersion in Toluol, Ø <0.1 mm
Sodium
Formel: Na
Dichte: 0,90 g/cm³
Flammpunkt: 4 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00085307
CAS-Nummer: 7440-23-5
UN: 3482
ADR: 4.3,I

Bestellen

Spezifikation Testergebnisse

Appearance Dispersion in toluene
Additional info Trace analysis based on 100% sodium metal purity
Strength ≥30 % (Na)
Chloride (Cl) ≤50 ppm
Calcium (Ca) ≤550 ppm
Potassium (K) ≤300 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR