2-Hydroxypyridin-N-oxid 97%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
243790250
243791000
243795000
ACRO243790250EA
67
EUR
ACRO243790250
ACRO243791000
ACRO243795000
2-Hydroxypyridin-N-oxid 97%
2-Hydroxypyridin-N-oxid
Formel:
C₅H₅NO₂ Schmelzpunkt: 147…152 °C Flammpunkt: 100 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00006195 CAS-Nummer: 13161-30-3 |
Spezifikation Testergebnisse
Appearance | Off-white to beige Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 147 to 152 °C |
Titration with NaOH | ≥96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...