Benzo-18-krone-6 (Benzo-1,4,7,10,13,16-hexoxacyclooctadecan) 97%

Lieferant: Thermo Fisher Scientific
Achtung

243980010 243980050
ACRO243980010EA 40.4 EUR
ACRO243980010 ACRO243980050
Benzo-18-krone-6 (Benzo-1,4,7,10,13,16-hexoxacyclooctadecan) 97%
Benzo-18-krone-6 (Benzo-1,4,7,10,13,16-hexoxacyclooctadecan)
Formel: C₁₆H₂₄O₆
Schmelzpunkt: 40…45 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00062741
CAS-Nummer: 14098-24-9

Bestellen

Spezifikation Testergebnisse

Appearance White to off-white Powder or crystals and/or chunks
Infrared spectrum Conforms
Melting point 40 to 45 °C
GC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR