2-Hydroxynicotinsäure 98%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 2-Hydroxypyridin-3-carbonsäure, 1,2-Dihydro-2-oxonicotinsäure, 2-Hydroxy-3-pyridincarbonsäure

247220250 247221000
ACRO247220250EA 44.8 EUR
ACRO247220250 ACRO247221000
2-Hydroxynicotinsäure 98%
2-Hydroxynicotinsäure
Formel: C₆H₅NO₃
Schmelzpunkt: 259…261 °C
MDL: MFCD00010100
CAS-Nummer: 609-71-2
EINECS: 210-198-2

Bestellen

Spezifikation Testergebnisse

Appearance White to cream to light yellow Powder
Infrared spectrum Conforms
Melting point 256 to 262 °C
Titration with NaOH ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR