meso-Hydrobenzoin 95%

Lieferant: Thermo Fisher Scientific

247680050 247680250
ACRO247680050EA 65.7 EUR
ACRO247680050 ACRO247680250
meso-Hydrobenzoin 95%
meso-Hydrobenzoin
Formel: C₁₄H₁₄O₂
Schmelzpunkt: 136…137 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00064253
CAS-Nummer: 579-43-1

Bestellen

Spezifikation Testergebnisse

Appearance White Crystalline powder and or lumps
Infrared spectrum Conforms
Melting point 134 to 140 °C
GC ≥94 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR