3-Amino-1H-1,2,4-triazole-5-carboxylic acid xhydrate 98%

Lieferant: Thermo Scientific
Achtung

Synonyme: 3-Amino-1,2,4-triazole-5-carboxylic acid xhydrate, 5-Amino-2H-1, 2, 4-triazole-3-carboxylic acid xhydrate

249301000
ACRO249301000EA 128 EUR
ACRO249301000
3-Amino-1H-1,2,4-triazole-5-carboxylic acid xhydrate 98%
3-Amino-1H-1,2,4-triazole-5-carboxylic acid xhydrate
Formel: C₃H₄N₄O₂·xH₂O
Molecular Weight: 128,09 g/mol (anhy)
Schmelzpunkt: 178…183 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00149595
CAS-Nummer: 1266615-63-7

Bestellen

Spezifikation Testergebnisse

Appearance Off-white Crystalline powder and/or chunks
Infrared spectrum Conforms
Melting point 178 to 183 °C
Loss on drying ≤2 %
Titration excess NaOH ≥97.5 % (On dry substance)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR