Methyl-2-brombenzoat 98%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
2-Brombenzoesäuremethylester
249810500
ACRO249810500EA
78.8
EUR
ACRO249810500
Methyl-2-brombenzoat 98%
Methyl-2-brombenzoat
Formel:
C₈H₇BrO₂ Molecular Weight: 215,05 g/mol Siedepunkt: 253 °C (1013 hPa) Dichte: 1,51 g/cm³ |
MDL:
MFCD00016328 CAS-Nummer: 610-94-6 EINECS: 210-241-5 |
Spezifikation Testergebnisse
Appearance | Clear light yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.5575 to 1.5595 (20°C, 589 nm) |
GC | ≥97.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...