L(+)-Calciumlactat Pentahydrat 98%
Lieferant: Thermo Scientific
Synonyme:
(S)-(+)-Milchsäure Calciumsalz Pentahydrat, (S)-(+)-2-Hydroxypropionsäure Calciumsalz Pentahydrat, Calcium-(S)-(+)-2-hydroxypropionat Pentahydrat, L(+)-Milchsäure Calciumsalz Pentahydrat, Calcium-L(+)-2-hydroxypropionat Pentahydrat
250200010
ACRO250200010EA
54.6
EUR
ACRO250200010
L(+)-Calciumlactat Pentahydrat 98%
L(+)-Calciumlactat Pentahydrat
Formel:
C₆H₁₀CaO₆·5H₂O Molecular Weight: 308,3 g/mol Schmelzpunkt: 240 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00150344 CAS-Nummer: 865717-20-0 EINECS: 248-953-3 |
Spezifikation Testergebnisse
Appearance | White Powder |
Infrared spectrum | Conforms |
Titration Complexometric | ≥97.5 % (on dry substance) |
Loss on drying | 22.0 % to 27.0 % |
Heavy metals | ≤10 ppm |
pH | 6.0 to 8.0 (5 % in water) |
Chloride (Cl) | ≤80 ppm |
Phosphate (PO4) | ≤150 ppm |
Sulfate (SO4) | ≤400 ppm |
Arsenic (As) | ≤1 ppm |
Calcium (Ca) | 13.4 to 14.5 % |
Iron (Fe) | ≤40 ppm |
Alkalies+Mg (as sulfates) | ≤1.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...