Isonicotinic anhydride 90% remainder picolinic and nicotinic acid-anhydride

Lieferant: Thermo Scientific
Gefahr

250280050
ACRO250280050EA 449 EUR
ACRO250280050
Isonicotinic anhydride 90% remainder picolinic and nicotinic acid-anhydride
Isonicotinic anhydride

Appearance: Off-white to beige to light brown Crystalline powder

Formel: C₁₂H₈N₂O₃
Molecular Weight: 228,207 g/mol
Schmelzpunkt: 133 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00062784
CAS-Nummer: 7082-71-5

Bestellen

Spezifikation Testergebnisse

Appearance Off-white to beige to light brown Crystalline powder
Infrared spectrum Conforms
Titration with HClO4 ≥88 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR