ɑ-Cyclopentyl-DL-mandelic acid 98%
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
alpha-Cyclopentylmandelic Acid, α-Cyclopentylmandelic acid
250970050
ACRO250970050EA
95.1
EUR
ACRO250970050
ɑ-Cyclopentyl-DL-mandelic acid 98%
alpha-Cyclopentyl-DL-mandelic acid
Appearance: White to beige Powder
Formel:
C₁₃H₁₆O₃ Molecular Weight: 220,27 g/mol Siedepunkt: 110 °C (,5 mmHg) Schmelzpunkt: 144…150 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00019296 CAS-Nummer: 427-49-6 UN: 3261 ADR: 8,III |
Spezifikation Testergebnisse
Appearance | White to beige Powder |
Infrared spectrum | Conforms |
Melting point | 144 to 150 °C |
Titration with NaOH | ≥97.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...