Ethyl-2-amino-4,5,6,7-tetrahydrobenzo[b]thiophen-3-carboxylat 99%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 2-Amino-4,5,6,7-tetrahydrobenzo[b]thiophene-3-carboxylic Acid Ethyl Ester, Ethyl 2-amino-4,5,6,7-tetrahydrobenzo[b]thiophene-3-carboxylate, 2-Amino-3-carbethoxy-4,5-tetramethylenethiophene, 2-Amino-3-ethoxycarbonyl-4,5-tetramethylenethiophene

251280100
ACRO251280100EA 63.5 EUR
ACRO251280100
Ethyl-2-amino-4,5,6,7-tetrahydrobenzo[b]thiophen-3-carboxylat 99%
Ethyl-2-Amino-4,5,6,7-tetrahydrobenzo[b]thiophene-3-carboxylate

Appearance: Yellow Crystalline powder

Formel: C₁₁H₁₅NO₂S
Schmelzpunkt: 113…117 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00005862
CAS-Nummer: 4506-71-2
UN: 2811
ADR: 6.1,III

Bestellen

Spezifikation Testergebnisse

Appearance Yellow Crystalline powder
Infrared spectrum Conforms
Melting point 114 to 117 °C
GC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR