D(-)-Cystein 99%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
(S)-(-)-Cystein, H-D-Cys-OH, (S)-(-)-2-Amino-3-mercaptopropansäure
254300010
ACRO254300010EA
198
EUR
ACRO254300010
D(-)-Cystein 99%
D(-)-Cystein
Formel:
C₃H₇NO₂S Schmelzpunkt: 230 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00066461 CAS-Nummer: 921-01-7 |
Spezifikation Testergebnisse
Appearance | White Crystalline powder |
Infrared spectrum | Conforms |
Titration Iodometric | ≥98.5 % |
Specific optical rotation | -8.0° to -9.5° (20°C, 589 nm) (C=5, 2 N HCl) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...