Promethazine hydrochloride

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 10-[2-(Dimethylamino)propyl]phenothiazine Hydrochloride, N,N-Dimethyl-1-(10H-phenothiazin-10-yl)propan-2-amine hydrochloride

254580050 254580250 254581000
ACRO254580050EA 39.3 EUR
ACRO254580050 ACRO254580250 ACRO254581000
Promethazine hydrochloride
Promethazine hydrochloride
Formel: C₁₇H₂₀N₂S
Molecular Weight: 320.88 g/mol
Schmelzpunkt: 227…230 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00012652
CAS-Nummer: 58-33-3
UN: 3077
ADR: 9 III

Bestellen

Spezifikation Testergebnisse

Appearance White to light yellow Powder
Infrared spectrum Conforms
Loss on drying ≤0.5 % (105°C, 4 hr)
Solubility (10% in water) Clear colorless to light yellow
Titration with HClO4 97.5 to 101.5 % (on dry substance)

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR