Quecksilber(II)acetat 99+%

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: Essigsäure Quecksilber(II)salz, Quecksilberdiacetat

255650050 255651000 255650010
ACRO255650050EA 39.7 EUR
ACRO255650050 ACRO255651000 ACRO255650010
Quecksilber(II)acetat 99+%
Quecksilber(II)acetat
Formel: (H₃CCOO)₂Hg
Schmelzpunkt: 179…182 °C
Dichte: 3,27 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00012165
CAS-Nummer: 1600-27-7
EINECS: 216-491-1
UN: 1629
ADR: 6.1,II
Merck Index: 13,05898

Bestellen

Spezifikation Testergebnisse

Appearance White to off-white Crystalline powder
Infrared spectrum Conforms
Titration Complexometric ≥99.0 %
Residue after ignition ≤0.02 % (substances not reducible with formic acid)
Chloride (Cl) ≤50 ppm
Nitrate (NO3) ≤50 ppm
Sulfate (SO4) ≤50 ppm
Iron (Fe) ≤10 ppm
Heavy metals (as Pb) ≤20 ppm
Mercurous Mercury (as Hg) ≤4000 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR