(±)-γ-Undecalacton 98%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: (±)-4-Hydroxyundecansäurelacton, (±)-Undecan-4-olid, (±)-γ-Undekalacton

259490250 259491000 259495000
ACRO259490250EA 23.4 EUR
ACRO259490250 ACRO259491000 ACRO259495000
(±)-γ-Undecalacton 98%
(±)-γ-Undecalacton
Formel: C₁₁H₂₀O₂
Molecular Weight: 184.28 g/mol
Siedepunkt: 286 °C (760 mmHg)
Dichte: 0.9425 g/cm³
Flammpunkt: 145 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00005405
CAS-Nummer: 104-67-6
EINECS: 203-225-4

Bestellen

Spezifikation Testergebnisse

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4490 to 1.4540 (20°C, 589 nm)
GC ≥97.5 %
Specific gravity (25°C) 0.939 to 0.948


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR