L-(-)-α-Amino-γ-butyrolactone hydrobromide 99%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: L-(-)-α-Amino-γ-butyrolactone hydrobromide, (S)-2-amino-4-butyrolactone hydrobromide, (S)-(-)-α-Amino-γ-butyrolactone hydrobromide, (S)-3-Aminodihydrofuran-2(3H)-one hydrobromide

259940010 259940050
ACRO259940010EA 37.2 EUR
ACRO259940010 ACRO259940050
L-(-)-α-Amino-γ-butyrolactone hydrobromide 99%
L-(-)-alpha-Amino-gamma-butyrolactone hydrobromide
Formel: C₄H₇NO₂·HBr
Molecular Weight: 182.017 g/mol
Schmelzpunkt: 236…238.9 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00674493
CAS-Nummer: 15295-77-9

Bestellen

Spezifikation Testergebnisse

Appearance White to light brown Powder
Infrared spectrum Conforms
Titration Argentometric ≥98.5 %
Specific optical rotation -20.5° to -22.5° (20°C, 589 nm) (c=4, H2O)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR