Hydroxychloroquin-sulfat 98%

Lieferant: Thermo Fisher Scientific
Achtung

263010250 263011000
ACRO263010250EA 115 EUR
ACRO263010250 ACRO263011000
Hydroxychloroquin-sulfat 98%
Hydroxychloroquine sulphate

CAS No.: 747-36-4

Formel: C₁₈H₂₆ClN₃O
Schmelzpunkt: 240 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00078203
CAS-Nummer: 747-36-4

Bestellen

Spezifikation Testergebnisse

Appearance White to off-white Crystalline powder
Infrared spectrum Conforms
Loss on drying ≤2 % (105°C, 2 h)
Heavy metals ≤20 ppm
Sulfated ash ≤0.2 %
UV spectrophotometry ≥97.5 % (on dried basis)
Chloride (Cl) ≤350 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR