trans-(+)-Chrysanthemic acid 99+%

Lieferant: Thermo Fisher Scientific
Achtung

263635000
ACRO263635000EA 223 EUR
ACRO263635000
trans-(+)-Chrysanthemic acid 99+%
trans-(+)-Chrysanthemic acid

Appearance: Clear colorless to light yellow Liquid

Formel: C₁₀H₁₆O₂
Schmelzpunkt: 17…21 °C
Lagertemperatur: Kühlschrank
MDL: MFCD01941572
CAS-Nummer: 4638-92-0

Bestellen

Spezifikation Testergebnisse

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4760 to 1.4790 (20°C, 589 nm)
GC ≥99.0 %
Specific optical rotation +13° to +15° (20°C, 589 nm) (c=2, ethanol)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR