(-)-ɑ-Methoxy-ɑ-(trifluoromethyl)phenylacetyl chloride 98+%

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: (-)-MTPA-Cl, (-)-Mosher's Acid Chloride, (R)-(-)-alpha-Methoxy-alpha-(trifluoromethyl)phenylacetyl chloride

264120010 264122500
ACRO264120010EA 347 EUR
ACRO264120010 ACRO264122500
(-)-ɑ-Methoxy-ɑ-(trifluoromethyl)phenylacetyl chloride 98+%
(-)-alpha-Methoxy-alpha-(trifluoromethyl)phenylacetyl chloride

CAS No.: 39637-99-5

Formel: C₁₀H₈ClF₃O₂
Molecular Weight: 252.62 g/mol
Siedepunkt: 213…214 °C (760 mmHg)
Dichte: 1.34 g/cm³
Flammpunkt: 89 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00044400
CAS-Nummer: 39637-99-5
UN: 3265
ADR: 8 II

Bestellen

Spezifikation Testergebnisse

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4675 to 1.4715 (20°C, 589 nm)
GC ≥98.0 %
Specific optical rotation -128° to -134° (20°C, 589 nm) (c=1, CHCl3)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR