Methylrot, rein Indikator

Lieferant: Thermo Fisher Scientific

Synonyme: 2-(4-(Dimethylamino)phenylazo)benzoesäure

264440250 264441000 264445000
ACRO264440250EA 38.3 EUR
ACRO264440250 ACRO264441000 ACRO264445000
Methylrot, rein Indikator
Methylrot
Formel: C₁₅H₁₅N₃O₂
Molecular Weight: 269.3 g/mol
Schmelzpunkt: 178…182 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00002425
CAS-Nummer: 493-52-7
EINECS: 207-776-1
Merck Index: 12,06199

Bestellen

Spezifikation Testergebnisse

Appearance Dark red to brown to purple to violet Powder or crystals
Infrared spectrum Conforms
Melting point 178 to 182 °C
Solubility (0.1 % in ethanol) Clear
Specific extinction E (1%/1cm) ≥650 (430 nm, in buffer)
Visual transition interval from pH 4.2 (pink or red) to pH 6.2 (yellow)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR