Magnesiumhydrogenorthophosphat Trihydrat 98%, reinst

Lieferant: Thermo Fisher Scientific

Synonyme: Magnesiumphosphat dibasisch Trihydrat, Magnesiumorthophosphat dibasisch Trihydrat, Magnesiumhydrogenorthophosphat Trihydrat

264780010
ACRO264780010EA 62.9 EUR
ACRO264780010
Magnesiumhydrogenorthophosphat Trihydrat 98%, reinst
Magnesiumhydrogenorthophosphat Trihydrat
Formel: HMgO₄P·3H₂O
Dichte: 2,13 g/cm³
Lagertemperatur: Raumtemperatur
MDL: MFCD00150390
CAS-Nummer: 7782-75-4
EINECS: 231-823-5

Bestellen

Spezifikation Testergebnisse

Appearance White Powder
Titration Complexometric ≥97.5 %
Chloride (Cl) ≤200 ppm
Sulfate (SO4) ≤300 ppm
Arsenic (As) ≤1 ppm
Iron (Fe) ≤50 ppm
Heavy metals (as Pb) ≤20 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR