Ethyl-trans-2-amino-1-cyclohexanecarboxylate hydrochloride 98%

Lieferant: Thermo Fisher Scientific

265650010
ACRO265650010EA 166 EUR
ACRO265650010
Ethyl-trans-2-amino-1-cyclohexanecarboxylate hydrochloride 98%
Ethyl-trans-2-amino-1-cyclohexanecarboxylate hydrochloride

CAS No.: 28250-14-8

Formel: C₉H₁₈ClNO₂
Schmelzpunkt: 152…156 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00144297
CAS-Nummer: 28250-14-8

Bestellen

Spezifikation Testergebnisse

Appearance White to off-white Granular powder
Infrared spectrum Conforms
Melting point 152 to 156 °C
Titration Argentometric ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR