3-Aminobicyclo[2.2.1]hept-5-ene-2-carboxylic acid 98%

Lieferant: Thermo Fisher Scientific

265690010
ACRO265690010EA 221 EUR
ACRO265690010
3-Aminobicyclo[2.2.1]hept-5-ene-2-carboxylic acid 98%
3-Aminobicyclo[2.2.1]hept-5-ene-2-carboxylic acid

Appearance: Beige to light grey Powder

Formel: C₈H₁₁NO₂
Schmelzpunkt: 261…263 °C
Lagertemperatur: Raumtemperatur
CAS-Nummer: 92511-32-5

Bestellen

Spezifikation Testergebnisse

Appearance Beige to light grey Powder
Infrared spectrum Conforms
Water ≤10.5 % (K.F.)
Titration with HClO4 ≥97.5 % (On dry substance)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR