cis-2-Hydroxymethyl-1-cyclohexylamine hydrochloride 99%
Lieferant: Thermo Scientific
265860010
ACRO265860010EA
206
EUR
ACRO265860010
cis-2-Hydroxymethyl-1-cyclohexylamine hydrochloride 99%
cis-2-Hydroxymethyl-1-cyclohexylamine hydrochloride
Appearance: White to light beige Crystalline powder
Formel:
C₇H₁₆ClNO Schmelzpunkt: 131…135 °C Lagertemperatur: Raumtemperatur |
CAS-Nummer:
5691-37-2 |
Spezifikation Testergebnisse
Appearance | White to light beige Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 131 to 135 °C |
Titration Argentometric | ≥98.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...