3-Methyl-5-methoxycarbonyl-4-piperidone hydrochloride 99%

Lieferant: Thermo Scientific

268085000
ACRO268085000EA 65 EUR
ACRO268085000
3-Methyl-5-methoxycarbonyl-4-piperidone hydrochloride 99%
3-Methyl-5-methoxycarbonyl-4-piperidone hydrochloride

Appearance: White Crystalline powder

Formel: C₈H₁₄ClNO₃
Schmelzpunkt: 160…175 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00143176
CAS-Nummer: 205692-64-4

Bestellen

Spezifikation Testergebnisse

Appearance White Crystalline powder
Infrared spectrum Conforms
Titration Argentometric ≥98.5 %
Titration with HClO4 ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR