Kaliumbromat 99.5% zur Analyse

Lieferant: Thermo Fisher Scientific
Gefahr

268390010 268392500
ACRO268390010EA 182 EUR
ACRO268390010 ACRO268392500
Kaliumbromat 99.5% zur Analyse
Potassium bromate
Formel: KBrO₃
Schmelzpunkt: 350 °C
Dichte: 3,27 g/cm³
Lagertemperatur: Raumtemperatur
MDL: MFCD00011359
CAS-Nummer: 7758-01-2
EINECS: 231-829-8
UN: 1484
ADR: 5.1,II
Merck Index: 12,07779

Bestellen

Spezifikation Testergebnisse

Appearance White Crystalline powder
Titration Iodometric ≥99.5 %
Loss on drying ≤0.1 % (105°C)
pH 5 to 9 (5 % at 20°C)
Bromide (Br) ≤500 ppm
Sulfate (SO4) ≤50 ppm
Iron (Fe) ≤5 ppm
Sodium (Na) ≤100 ppm
Heavy metals (as Pb) ≤5 ppm
Total nitrogen compounds ≤0.001 % (as N)
Insoluble matter ≤0.005 % (in water)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR