Rhodium(II)acetat dimer (∼46% Rh), wasserfrei

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: Dirhodium tetraacetate, Tetrakis(acetato)dirhodium(II)

268630010 268632500 268630050
ACRO268630010EA 1152 EUR
ACRO268630010 ACRO268632500 ACRO268630050
Rhodium(II)acetat dimer (∼46% Rh), wasserfrei
Rhodium(II)acetat dimer
Formel: C₈H₁₂O₈Rh₂
Molecular Weight: 441.987 g/mol
Schmelzpunkt: 205 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00003538
CAS-Nummer: 15956-28-2
EINECS: 240-084-8

Bestellen

Spezifikation Testergebnisse

Appearance Green Crystalline powder
Infrared spectrum Conforms
Assay 46 % (Rh)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR