Tetrabutylammoniumdihydrogentrifluorid 50-55% (w/w) in 1,2-Dichloroethan
Lieferant: Thermo Scientific
|
Gefahr
|
268780025
ACRO268780025EA
88.5
EUR
ACRO268780025
Tetrabutylammoniumdihydrogentrifluorid 50-55% (w/w) in 1,2-Dichloroethan
Tetrabutylammoniumdihydrogentrifluorid
Formel:
C₁₆H₃₈F₃N Flammpunkt: 13 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00145365 CAS-Nummer: 99337-56-1 UN: 2924 ADR: 3,II |
Spezifikation Testergebnisse
Appearance | Clear light yellow to brown Solution |
Infrared spectrum | Conforms |
NMR | Consistent with structure |
Strength | 50 to 55 % (NMR) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...