1,3,6-Naphthalenetrisulphonic acid, sodium salt hydrate Gemisch aus Isomeren

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 1,3,6-Naphthalenetrisulphonic acid, xsodium salt, Sodium-3,6-disulphonaphthalene-1-sulphonate

269010010
ACRO269010010EA 158 EUR
ACRO269010010
1,3,6-Naphthalenetrisulphonic acid, sodium salt hydrate Gemisch aus Isomeren
1,3,6-Naphthalenetrisulphonic acid, sodium salt hydrate

CAS No.:

Formel: C₁₀H₇NaO₉S₃·xH₂O
Molecular Weight: 390.34 g/mol (anhy)
Lagertemperatur: Raumtemperatur
CAS-Nummer: 19437-42-4

Bestellen

Spezifikation Testergebnisse

Appearance White to yellow to beige Powder
Infrared spectrum Conforms
Loss on drying ≤12 %
HPLC ≥90 % (mix of isomers)
Sodium sulfate (Na2SO4) ≤5


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR