Kaliummethylmalonat 98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
Potassium 3-methoxy-3-oxopropanoate, Monomethyl monopotassium malonate, Monomethyl malonate potassium salt
269550250
269551000
269555000
ACRO269550250EA
40.8
EUR
ACRO269550250
ACRO269551000
ACRO269555000
Kaliummethylmalonat 98%
Kaliummethylmalonat
Formel:
C₄H₅KO₄ Molecular Weight: 156.18 g/mol Schmelzpunkt: >200 °C (Zers.) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00014021 CAS-Nummer: 38330-80-2 EINECS: 253-886-8 |
Spezifikation Testergebnisse
Appearance | White Crystalline powder |
Infrared spectrum | Conforms |
Titration with HClO4 | ≥97.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...