Hexafluoraceton Sesquihydrat 98%
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
1,1,1,3,3,3-Hexafluoroacetone sesquihydrate
269970250
ACRO269970250EA
151
EUR
ACRO269970250
Hexafluoraceton Sesquihydrat 98%
Hexafluoraceton Sesquihydrat
Formel:
C₃F₆O·1.5H₂O Molecular Weight: 193,05 g/mol Schmelzpunkt: 18…21 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00149063 CAS-Nummer: 13098-39-0 EINECS: 211-676-3 UN: 2552 ADR: 6.1,II |
Spezifikation Testergebnisse
Appearance | Clear light yellow to amber Liquid |
Infrared spectrum | Conforms |
Assay | ≥97.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...