Hydroxylammoniumchlorid 99+%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
270100010
270101000
270102500
270100050
ACRO270100010EA
197
EUR
ACRO270100010
ACRO270101000
ACRO270102500
ACRO270100050
Hydroxylammoniumchlorid 99+%
Hydroxylammoniumchlorid
Formel:
NH₂OH·HCl Siedepunkt: 306 °C (1013 hPa) Dichte: 1,67 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00051089 CAS-Nummer: 5470-11-1 EINECS: 226-798-2 UN: 2923 ADR: 8,III Merck Index: 13,04853 |
Spezifikation Testergebnisse
Appearance | White Crystalline powder or crystals and/or chunks |
Infrared spectrum | Conforms |
Titration with NaOH | 99.0 to 101.0 % |
Sulfate (SO4) | ≤0.005 % |
Iron (Fe) | ≤0.001 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...