Magnesiumacetat Tetrahydrat 99.5% zur Analyse

Lieferant: Thermo Fisher Scientific

Synonyme: Essigsäure Magnesiumsalz-Tetrahydrat, Magnesiumdi(acetat) Tetrahydrat, Essigsäure Magnesiumsalz Tetrahydrat, Magnesiumdi(acetat)-Tetrahydrat

270500010 270500050 270500051
ACRO270500010EA 128 EUR
ACRO270500010 ACRO270500050 ACRO270500051
Magnesiumacetat Tetrahydrat 99.5% zur Analyse
Magnesiumacetat Tetrahydrat
Formel: (H₃CCOO)₂Mg·4H₂O
Schmelzpunkt: 80 °C
Dichte: 1,454 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00149214
CAS-Nummer: 16674-78-5
EINECS: 205-554-9
Merck Index: 13,05676

Bestellen

Spezifikation Testergebnisse

Appearance White Crystalline powder
Infrared spectrum Conforms
Titration Complexometric ≥99.4 %
pH 7 to 9 (5% aq. soln. at 20°C)
Chloride (Cl) ≤10 ppm
Sulfate (SO4) ≤50 ppm
Barium (Ba) ≤10 ppm
Calcium (Ca) ≤100 ppm
Copper (Cu) ≤5 ppm
Iron (Fe) ≤5 ppm
Strontium (Sr) ≤50 ppm
Heavy metals (as Pb) ≤5 ppm
Total nitrogen compounds ≤0.001 % (as N)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR