Salpetersäure rauchend 90+% ACS

Lieferant: Thermo Scientific
Gefahr

Synonyme: Nitric acid solution

270620010
ACRO270620010EA 270 EUR
ACRO270620010
Salpetersäure rauchend 90+% ACS
Salpetersäure
Formel: HNO₃
Molecular Weight: 63,01 g/mol
Siedepunkt: 84 °C (760 mmHg)
Schmelzpunkt: –40 °C
Dichte: 1,511 g/cm³
Lagertemperatur: Raumtemperatur
MDL: MFCD00011349
CAS-Nummer: 7697-37-2
EINECS: 231-714-2
UN: 2032
ADR: 8,I
REACH: 01-2119487297-23
Merck Index: 13,06610

Bestellen

Spezifikation Testergebnisse

Appearance Clear colorless to yellow Liquid
Residue after ignition ≤0.002 %
Titration with NaOH ≥90.0 %
Dilution test Passes test
Chloride (Cl) ≤0.7 ppm
Sulfate (SO4) ≤5 ppm
Arsenic (As) ≤0.3 ppm
Iron (Fe) ≤2 ppm
Heavy metals (as Pb) ≤5 ppm
Dissolved oxides ≤0.1 % (as N2O3)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR