Triethylammoniumchlorid 99+%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
Triethylamin Hydrochlorid
270670010
270672500
ACRO270670010EA
103
EUR
ACRO270670010
ACRO270672500
Triethylammoniumchlorid 99+%
Triethylammonium chloride
Formel:
C₆H₁₆ClN Molecular Weight: 137.65 g/mol Schmelzpunkt: 254…260 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00012500 CAS-Nummer: 554-68-7 EINECS: 209-067-2 |
Spezifikation Testergebnisse
Appearance | White to off-white Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 254 to 260 °C |
Loss on drying | ≤1.0 % |
Solubility | (10 % in water) Clear colorless |
pH | 4.5 to 6.5 (1 % w/v solution) |
Titration with HClO4 | 99.0 to 101.0 % (On anhydrous substance) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...