(Benzotriazol-1-yloxy)tri(1-pyrrolidinyl)phosphoniumhexafluorophosphat (PyBOP) 99%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: PyBOP, ((1H-Benzo[d][1,2,3]triazol-1-yl)oxy)tri(pyrrolidin-1-yl)phosphonium hexafluorophosphate(V), 1H-Benzotriazol-1-yloxytri(1-pyrrolidinyl)phosphonium hexafluorophosphate

270720050 270720250
ACRO270720050EA 122 EUR
ACRO270720050 ACRO270720250
(Benzotriazol-1-yloxy)tri(1-pyrrolidinyl)phosphoniumhexafluorophosphat (PyBOP) 99%
(Benzotriazol-1-yloxy)tri(1-pyrrolidinyl)phosphoniumhexafluorophosphat (PyBOP)
Formel: C₁₈H₂₈F₆N₆OP₂
Schmelzpunkt: 145…160 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00077411
CAS-Nummer: 128625-52-5

Bestellen

Spezifikation Testergebnisse

Appearance White to off-white Crystalline powder
Infrared spectrum Conforms
Melting point 145 to 160 °C (Dec)
HPLC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR