Methyl-β-D-glucopyranosid Hemihydrat 99%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
Methyl beta-D-Glucoside
270920250
ACRO270920250EA
287
EUR
ACRO270920250
Methyl-β-D-glucopyranosid Hemihydrat 99%
Methyl-β-D-glucopyranosid Hemihydrat
Formel:
C₇H₁₄O₆·0.5H₂O Molecular Weight: 203,19 g/mol Schmelzpunkt: 111…113 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00149442 CAS-Nummer: 7000-27-3 EINECS: 211-909-9 |
Spezifikation Testergebnisse
Appearance | White Crystalline powder |
Infrared spectrum | Conforms |
Water | ≤5.4 % (K.F.) |
Solubility | (10 % in water) Clear colorless |
HPLC | ≥98.5 % (ELSD) |
Specific optical rotation | -31° to -35° (20°C, 589 nm) (c=4, water) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...