1,2:5,6-Di-O-isopropylidene-α-D-allofuranose 98%

Lieferant: Thermo Fisher Scientific

270960100
ACRO270960100EA 275 EUR
ACRO270960100
1,2:5,6-Di-O-isopropylidene-α-D-allofuranose 98%
1,2:5,6-Di-O-isopropylidene-alpha-D-allofuranose

CAS No.: 2595-05-3

Formel: C₁₂H₂₀O₆
Schmelzpunkt: 73…77 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00135634
CAS-Nummer: 2595-05-3

Bestellen

Spezifikation Testergebnisse

Appearance White to off-white Crystalline powder or crystals and/or chunks
Infrared spectrum Conforms
Melting point 72 to 77 °C
GC ≥97.5 %
Specific optical rotation +33.0° to +39.0° (20 °C, 589 nm) (c=1, CHCl3)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR